Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rbm39
Entrez Gene ID
170791
Gene Name
RNA binding motif protein 39
Gene Aliases
1500012C14Rik, 2310040E03Rik, B330012G18Rik, C79248, R75070, Rnpc2, caper
Chromosome Location
Chr.2: 156147239 - 156180238 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK009719.1 - 2 - 3 333 65
AK156737.1 - 2 - 3 329 65
AK178368.1 - 1 - 2 75 65
AK178834.1 - 1 - 2 75 65

View Full Product Details