Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Limd2
Entrez Gene ID
67803
Gene Name
LIM domain containing 2
Gene Aliases
0610025L06Rik, AI413966
Chromosome Location
Chr.11: 106156256 - 106161039 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK171794.1 - 1 - 2 373 68

View Full Product Details