Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lpcat2
Entrez Gene ID
270084
Gene Name
lysophosphatidylcholine acyltransferase 2
Gene Aliases
A330042H22, Aytl1, Aytl1a, lpafat1
Chromosome Location
Chr.8: 92855330 - 92919279 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC144806.1 - 2 - 3 366 59
BC150666.1 - 2 - 3 366 59

View Full Product Details