The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256033.1 | NP_001242962.1 | 10 - 11 | 1094 | 92 |
GenBank mRNA | AK122429.1 | - | 5 - 6 | 385 | 92 |
BC138373.1 | - | 10 - 11 | 1066 | 92 |