The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081164.1 | NP_001074633.1 | 3 - 4 | 524 | 117 |
NM_001256033.1 | NP_001242962.1 | 3 - 4 | 524 | 117 | |
GenBank mRNA | AW494258.1 | - | 3 - 4 | 524 | 117 |
BC138373.1 | - | 3 - 4 | 496 | 117 | |
BC145259.1 | - | 3 - 4 | 496 | 117 |