Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cse1l
Entrez Gene ID
110750
Gene Name
chromosome segregation 1-like (S. cerevisiae)
Gene Aliases
2610100P18Rik, AA407533, Capts, Cas, Xpo2
Chromosome Location
Chr.2: 166906051 - 166946389 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023565.3 NP_076054.1 17 - 18 1904 120
GenBank mRNA AF301152.1 - 17 - 18 1904 120
AK138940.1 - 3 - 4 328 120
BC145393.1 - 16 - 17 1675 120

View Full Product Details