The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010484.2 | NP_034614.2 | 13 - 14 | 2000 | 105 |
GenBank mRNA | AF013604.1 | - | 12 - 13 | 1813 | 105 |
AK008410.1 | - | 2 - 3 | 105 | ||
AY765218.1 | - | 2 - 3 | 258 | 105 | |
BC108978.1 | - | 13 - 14 | 1982 | 105 | |
BC108979.1 | - | 13 - 14 | 1982 | 105 | |
X66119.1 | - | 12 - 13 | 1594 | 105 |