The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083935.1 | NP_001077404.1 | 11 - 12 | 1390 | 66 |
NM_001083936.1 | NP_001077405.1 | 10 - 11 | 1423 | 66 | |
NM_011199.2 | NP_035329.2 | 12 - 13 | 1392 | 66 | |
GenBank mRNA | BC013446.1 | - | 12 - 13 | 1369 | 66 |
BC051981.1 | - | 10 - 11 | 1294 | 66 | |
X78936.1 | - | 10 - 11 | 1169 | 66 |