The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109873.1 | NP_001103343.1 | 3 - 4 | 545 | 65 |
NM_009824.2 | NP_033954.2 | 3 - 4 | 526 | 65 | |
GenBank mRNA | AF038029.1 | - | 3 - 4 | 408 | 65 |
BB658300.1 | - | 3 - 4 | 288 | 65 | |
BC141396.1 | - | 3 - 4 | 355 | 65 | |
BC151156.1 | - | 3 - 4 | 355 | 65 |