Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prkaca
Entrez Gene ID
18747
Gene Name
protein kinase, cAMP dependent, catalytic, alpha
Gene Aliases
PKCD, Pkaca
Chromosome Location
Chr.8: 83972978 - 83996444 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277898.1 NP_001264827.1 2 - 3 273 101
NM_008854.5 NP_032880.1 2 - 3 316 101
GenBank mRNA AF239743.1 - 2 - 3 273 101
BC003238.1 - 2 - 3 306 101
BC054834.1 - 2 - 3 301 101
BU059097.1 - 2 - 3 266 101
JN566056.1 - 2 - 3 230 101

View Full Product Details