Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsen54
Entrez Gene ID
76265
Gene Name
tRNA splicing endonuclease 54 homolog (S. cerevisiae)
Gene Aliases
0610034P02Rik
Chromosome Location
Chr.11: 115813743 - 115823102 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029557.1 NP_083833.1 3 - 4 307 57
GenBank mRNA AK002758.1 - 3 - 4 292 57
AK089005.1 - 3 - 4 274 57
BC126953.1 - 2 - 3 431 57
BY203746.1 - 3 - 4 312 57

View Full Product Details