The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102611.1 | NP_001096081.1 | 2 - 3 | 458 | 98 |
GenBank mRNA | AK030380.1 | - | 2 - 3 | 296 | 98 |
AK089959.1 | - | 2 - 3 | 394 | 98 | |
AK141484.1 | - | 2 - 3 | 371 | 98 | |
AK166057.1 | - | 2 - 3 | 396 | 98 | |
BC049810.1 | - | 2 - 3 | 320 | 98 | |
CN693729.1 | - | 2 - 3 | 458 | 98 |