Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab4a
Entrez Gene ID
19341
Gene Name
RAB4A, member RAS oncogene family
Gene Aliases
AI848268, Rab4
Chromosome Location
Chr.8: 123805847 - 123835291 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009003.3 NP_033029.2 3 - 4 366 126
GenBank mRNA AB232591.1 - 3 - 4 226 126
AK136566.1 - 3 - 4 355 126
AK145452.1 - 3 - 4 126
AK161877.1 - 3 - 4 334 126
BC138508.1 - 3 - 4 279 126
CO806247.1 - - 382 126

View Full Product Details