The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026181.1 | NP_080457.1 | 13 - 14 | 1919 | 79 |
GenBank mRNA | AK004868.1 | - | 13 - 14 | 1919 | 79 |
AK014367.1 | - | 13 - 14 | 1898 | 79 | |
AK154571.1 | - | 13 - 14 | 1900 | 79 | |
BC026394.1 | - | 7 - 8 | 1085 | 79 | |
BC100577.1 | - | 13 - 14 | 1894 | 79 | |
BC125584.1 | - | 13 - 14 | 1910 | 79 | |
BC132155.1 | - | 13 - 14 | 1910 | 79 |