The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013799.1 | NP_001013821.1 | 4 - 5 | 462 | 60 |
GenBank mRNA | AK041301.1 | - | 4 - 5 | 462 | 60 |
BC147328.1 | - | 4 - 5 | 400 | 60 | |
BC147329.1 | - | 4 - 5 | 400 | 60 |