The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190984.1 | NP_001177913.1 | 3 - 4 | 281 | 58 |
NM_001190985.1 | NP_001177914.1 | 2 - 3 | 499 | 58 | |
NM_021295.3 | NP_067270.1 | 3 - 4 | 315 | 58 | |
GenBank mRNA | AJ294535.1 | - | 3 - 4 | 308 | 58 |
AK146120.1 | - | 2 - 3 | 491 | 58 | |
AK182687.1 | - | 2 - 3 | 209 | 58 | |
AK186065.1 | - | 2 - 3 | 210 | 58 | |
AK210427.1 | - | 2 - 3 | 209 | 58 | |
BC058560.1 | - | 3 - 4 | 281 | 58 | |
Y11550.1 | - | 3 - 4 | 280 | 58 | |
Y16518.1 | - | 2 - 3 | 459 | 58 |