Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fchsd1
Entrez Gene ID
319262
Gene Name
FCH and double SH3 domains 1
Gene Aliases
A030002D08Rik
Chromosome Location
Chr.18: 37957434 - 37969814 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175684.4 NP_783615.2 8 - 9 700 71
GenBank mRNA AK079461.1 - 2 - 3 172 71
BC052886.1 - 8 - 9 731 71
BC057367.1 - 8 - 9 700 71

View Full Product Details