Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cntln
Entrez Gene ID
338349
Gene Name
centlein, centrosomal protein
Gene Aliases
AI448092, B430108F07Rik, D530005L17Rik
Chromosome Location
Chr.4: 84883512 - 85131923 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175275.4 NP_780484.2 4 - 5 825 126
NM_177385.4 NP_796359.1 4 - 5 825 126
GenBank mRNA AK031604.1 - 4 - 5 731 126
AK052558.1 - 4 - 5 745 126
AK165111.1 - 3 - 4 298 126
BC125622.1 - 4 - 5 700 126
BC125624.1 - 4 - 5 700 126

View Full Product Details