Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scp2
Entrez Gene ID
20280
Gene Name
sterol carrier protein 2, liver
Gene Aliases
AA409774, AA409893, C76618, C79031, NSL-TP, SCP-2, SCP-X, SCPX, ns-LTP
Chromosome Location
Chr.4: 108043830 - 108144971 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011327.4 NP_035457.1 6 - 7 624 121
GenBank mRNA AK004860.2 - 6 - 7 554 121
BC018384.1 - 6 - 7 579 121
BC034613.1 - 6 - 7 545 121
M91458.1 - 6 - 7 539 121

View Full Product Details