Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Abcb6
Entrez Gene ID
74104
Gene Name
ATP-binding cassette, sub-family B (MDR/TAP), member 6
Gene Aliases
1200005B17Rik
Chromosome Location
Chr.1: 75171640 - 75180283 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023732.2 NP_076221.1 12 - 13 1973 76
GenBank mRNA AK004605.1 - 12 - 13 2053 76
AK151165.1 - 12 - 13 2082 76
AK164003.1 - 12 - 13 1977 76
AK168642.1 - 12 - 13 2074 76
BC006634.1 - 12 - 13 1973 76

View Full Product Details