Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mbp
Entrez Gene ID
17196
Gene Name
myelin basic protein
Gene Aliases
C76307, Hmbpr, R75289, golli-mbp, mld, shi
Chromosome Location
Chr.18: 82475123 - 82585637 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025251.2 NP_001020422.1 6 - 7 659 57
NM_001025254.2 NP_001020425.1 5 - 6 581 57
NM_001025255.2 NP_001020426.1 5 - 6 581 57
NM_001025258.2 NP_001020429.1 4 - 5 548 57
GenBank mRNA AK179209.1 - 3 - 4 205 57
AK184274.1 - 3 - 4 203 57
AK208189.1 - 4 - 5 236 57
BC004704.1 - 5 - 6 543 57
CK623660.1 - 5 - 6 502 57
M15062.1 - 4 - 5 406 57

View Full Product Details