Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nhej1
Entrez Gene ID
75570
Gene Name
nonhomologous end-joining factor 1
Gene Aliases
1700029B21Rik, XLF, cernunnos
Chromosome Location
Chr.1: 74967346 - 75046653 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029342.4 NP_083618.3 - 94 64
GenBank mRNA BC006063.1 - - 68 64

View Full Product Details