Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sncaip
Entrez Gene ID
67847
Gene Name
synuclein, alpha interacting protein (synphilin)
Gene Aliases
2810407O15Rik, 4933427B05Rik, BB104594, SYPH1
Chromosome Location
Chr.18: 52767811 - 52915935 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199151.1 NP_001186080.1 6 - 7 1478 69
NM_001199153.1 NP_001186082.1 6 - 7 1383 69
NM_001199154.1 NP_001186083.1 6 - 7 1653 69
NM_026408.4 NP_080684.3 6 - 7 1478 69
GenBank mRNA AF348447.1 - 6 - 7 1406 69
AK013040.1 - 4 - 5 1212 69
AK017012.1 - 3 - 4 413 69
AK044974.1 - 6 - 7 1676 69
AK081942.1 - 6 - 7 1657 69
AK082079.1 - 6 - 7 1482 69
AK132346.1 - 6 - 7 1581 69
AK160111.1 - 6 - 7 1482 69
BC098489.1 - 6 - 7 1382 69
BC130225.1 - 6 - 7 1368 69
BC152544.1 - 6 - 7 1366 69

View Full Product Details