Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Chst9
- Entrez Gene ID
-
71367
- Gene Name
- carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9
- Gene Aliases
-
5430438D01Rik
- Chromosome Location
-
Chr.18: 15451773 - 15760415 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK017407.1 |
- |
1 - 2 |
180 |
110 |
View Full Product Details