The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110516.1 | NP_001103986.1 | 8 - 9 | 1371 | 89 |
GenBank mRNA | AK149268.1 | - | 8 - 9 | 1371 | 89 |
AK161035.1 | - | 7 - 8 | 1385 | 89 | |
AY458844.1 | - | 7 - 8 | 1188 | 89 |