Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sos2
Entrez Gene ID
20663
Gene Name
son of sevenless homolog 2 (Drosophila)
Gene Aliases
SOS-2, mSOS-2
Chromosome Location
Chr.12: 69583761 - 69681852 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135559.1 NP_001129031.1 19 - 20 3329 77
GenBank mRNA Z11664.1 - 18 - 19 2973 77

View Full Product Details