Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tectb
Entrez Gene ID
21684
Gene Name
tectorin beta
Gene Aliases
Tctnb
Chromosome Location
Chr.19: 55180733 - 55196313 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009348.3 NP_033374.2 1 - 2 211 109
GenBank mRNA BC138888.1 - 1 - 2 140 109
BC138889.1 - 1 - 2 140 109
X99806.2 - 1 - 2 211 109

View Full Product Details