The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199151.1 | NP_001186080.1 | 11 - 12 | 3003 | 64 |
NM_001199153.1 | NP_001186082.1 | 11 - 12 | 2908 | 64 | |
GenBank mRNA | AK017012.1 | - | 8 - 9 | 1938 | 64 |
AK160111.1 | - | 11 - 12 | 3007 | 64 | |
BC098489.1 | - | 11 - 12 | 2907 | 64 | |
BC130225.1 | - | 11 - 12 | 2893 | 64 | |
BC152544.1 | - | 11 - 12 | 2891 | 64 |