The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010777.3 | NP_034907.1 | 6 - 7 | 1011 | 117 |
GenBank mRNA | L07507.1 | - | 6 - 7 | 971 | 117 |
L07509.1 | - | 5 - 6 | 333 | 117 |