Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hes2
Entrez Gene ID
15206
Gene Name
hairy and enhancer of split 2 (Drosophila)
Gene Aliases
HES-2
Chromosome Location
Chr.4: 152158867 - 152162469 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008236.4 NP_032262.2 2 - 3 170 73
GenBank mRNA AK053792.1 - 2 - 3 171 73
BC138112.1 - 2 - 3 498 73

View Full Product Details