The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026395.1 | NP_080671.1 | 5 - 6 | 469 | 91 |
GenBank mRNA | AK002550.1 | - | 5 - 6 | 469 | 91 |
AK003682.1 | - | 5 - 6 | 491 | 91 | |
AK013262.1 | - | 5 - 6 | 526 | 91 | |
AK078396.1 | - | 5 - 6 | 532 | 91 | |
AK168192.1 | - | 6 - 7 | 688 | 91 | |
BC029189.1 | - | 4 - 5 | 375 | 91 |