Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Scin
Entrez Gene ID
20259
Gene Name
scinderin
Gene Aliases
AW545522, adseverin
Chromosome Location
Chr.12: 40059769 - 40134228 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146196.1 NP_001139668.1 15 - 16 2138 93
NM_009132.2 NP_033158.2 13 - 14 1838 93
GenBank mRNA AK133484.1 - 15 - 16 2094 93
AK136080.1 - 13 - 14 1680 93
AK167739.1 - 15 - 16 2093 93
AK173295.1 - 15 - 16 2100 93
BC063328.1 - 15 - 16 2093 93
U04354.1 - 15 - 16 2093 93
Y13971.1 - 13 - 14 1793 93

View Full Product Details