The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177772.1 | NP_001171243.1 | 1 - 2 | 353 | 124 |
NM_021471.2 | NP_067446.1 | 1 - 2 | 353 | 124 | |
GenBank mRNA | AK045782.1 | - | - | 356 | 124 |
AK048926.1 | - | 1 - 2 | 323 | 124 | |
AK161010.1 | - | - | 323 | 124 | |
AY007379.1 | - | 1 - 2 | 189 | 124 | |
BC078456.1 | - | 1 - 2 | 303 | 124 |