The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033455.2 | NP_001028627.1 | 11 - 12 | 1875 | 93 |
GenBank mRNA | AK133459.1 | - | 11 - 12 | 1876 | 93 |
BC150930.1 | - | 11 - 12 | 1858 | 93 | |
BC151167.1 | - | 11 - 12 | 1858 | 93 |