Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Meis3
Entrez Gene ID
17537
Gene Name
Meis homeobox 3
Gene Aliases
AI573393, Mrg2
Chromosome Location
Chr.7: 16174849 - 16186510 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK083907.1 - 3 - 4 528 69
AK134758.1 - 3 - 4 506 69
AK155364.1 - 3 - 4 518 69
BC003762.1 - 3 - 4 509 69
BC117532.1 - 3 - 4 430 69
BC118513.1 - 3 - 4 430 69
U57344.1 - 3 - 4 485 69

View Full Product Details