Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tyrp1
Entrez Gene ID
22178
Gene Name
tyrosinase-related protein 1
Gene Aliases
Oca3, TRP-1, TRP1, Tyrp, b, brown, isa
Chromosome Location
Chr.4: 80834208 - 80851736 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282014.1 NP_001268943.1 6 - 7 1538 75
NM_001282015.1 NP_001268944.1 6 - 7 1532 75
NM_031202.3 NP_112479.1 6 - 7 1553 75
GenBank mRNA AK148332.1 - 6 - 7 1538 75
AK148370.1 - 6 - 7 1444 75
AK148441.1 - 6 - 7 1444 75
AK177043.1 - 1 - 2 39 75
AK178803.1 - 1 - 2 39 75
AK188091.1 - 1 - 2 39 75
AK189488.1 - 1 - 2 39 75
AK192440.1 - 1 - 2 39 75
AK193585.1 - 1 - 2 39 75
AK194427.1 - 1 - 2 39 75
AK197740.1 - 1 - 2 39 75
AK198835.1 - 1 - 2 39 75
AK199149.1 - 1 - 2 39 75
AK199163.1 - 1 - 2 39 75
AK199776.1 - 1 - 2 39 75
AK200013.1 - 1 - 2 39 75
AK200365.1 - 1 - 2 75
AK200395.1 - 1 - 2 39 75
BC076598.1 - 6 - 7 1536 75
X03687.1 - 6 - 7 1438 75

View Full Product Details