The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199016.2 | NP_950181.2 | 3 - 4 | 1195 | 72 |
GenBank mRNA | AK090098.1 | - | 3 - 4 | 1196 | 72 |
AK157622.1 | - | 3 - 4 | 1091 | 72 | |
AK173048.1 | - | 4 - 5 | 1155 | 72 | |
BC027749.1 | - | 3 - 4 | 1149 | 72 |