Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Treml4
Entrez Gene ID
224840
Gene Name
triggering receptor expressed on myeloid cells-like 4
Gene Aliases
5031403H21Rik, BB137214, IDCP1, TLT-4, TLT4, Treml3
Chromosome Location
Chr.17: 48264191 - 48275607 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163795.1 NP_001157267.1 1 - 2 144 95
NM_001163796.1 NP_001157268.1 1 - 2 144 95
NM_172623.2 NP_766211.2 1 - 2 144 95
GenBank mRNA AK077228.1 - 1 - 2 144 95
AK156085.1 - 1 - 2 147 95
AY465530.1 - 1 - 2 102 95
AY465531.1 - 1 - 2 102 95
AY465532.1 - 1 - 2 102 95
BC117091.1 - 1 - 2 119 95
BC137666.1 - 1 - 2 119 95
DQ087185.1 - 1 - 2 66 95
DQ087186.1 - 1 - 2 66 95
DQ186655.1 - 1 - 2 98 95

View Full Product Details