Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rhag
Entrez Gene ID
19743
Gene Name
Rhesus blood group-associated A glycoprotein
Gene Aliases
CD241, Rh50, Rh50A
Chromosome Location
Chr.17: 40811099 - 40840754 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011269.2 NP_035399.1 1 - 2 217 93
GenBank mRNA AB015192.1 - 1 - 2 190 93
AF057526.1 - 1 - 2 217 93
AF065395.1 - 1 - 2 190 93
AK143851.1 - 1 - 2 218 93
AK157847.1 - 1 - 2 220 93
BC101941.1 - 1 - 2 214 93
BC101942.1 - 1 - 2 214 93
BC103512.1 - 1 - 2 214 93
BC103662.1 - 1 - 2 214 93

View Full Product Details