Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rhag
Entrez Gene ID
19743
Gene Name
Rhesus blood group-associated A glycoprotein
Gene Aliases
CD241, Rh50, Rh50A
Chromosome Location
Chr.17: 40811099 - 40840754 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011269.2 NP_035399.1 7 - 8 1121 61
GenBank mRNA AB015192.1 - 7 - 8 1094 61
AF057526.1 - 7 - 8 1121 61
AF065395.1 - 7 - 8 1094 61
AK143851.1 - 7 - 8 1122 61
AK157847.1 - 7 - 8 1124 61
BC101941.1 - 7 - 8 1118 61
BC101942.1 - 7 - 8 1118 61
BC103512.1 - 7 - 8 1118 61
BC103662.1 - 7 - 8 1118 61

View Full Product Details