Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Jag1
Entrez Gene ID
16449
Gene Name
jagged 1
Gene Aliases
ABE2, Gsfabe2, Htu, Ozz, Ser-1
Chromosome Location
Chr.2: 137081451 - 137116520 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013822.5 NP_038850.1 9 - 10 1490 101
GenBank mRNA AF171092.1 - 9 - 10 1495 101
AK137092.1 - 9 - 10 1390 101
BC058675.1 - 9 - 10 1302 101

View Full Product Details