Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Esf1
Entrez Gene ID
66580
Gene Name
ESF1, nucleolar pre-rRNA processing protein, homolog (S. cerevisiae)
Gene Aliases
2610101J03Rik, ABTAP, AW545818, C79684
Chromosome Location
Chr.2: 140119880 - 140170558 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081090.1 NP_001074559.1 10 - 11 2057 104
GenBank mRNA AK132230.1 - 9 - 10 1979 104
BC096029.1 - 10 - 11 2054 104
BC150893.1 - 10 - 11 2023 104
BC157979.1 - 10 - 11 2023 104

View Full Product Details