Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Esf1
Entrez Gene ID
66580
Gene Name
ESF1, nucleolar pre-rRNA processing protein, homolog (S. cerevisiae)
Gene Aliases
2610101J03Rik, ABTAP, AW545818, C79684
Chromosome Location
Chr.2: 140119880 - 140170558 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081090.1 NP_001074559.1 6 - 7 1494 89
GenBank mRNA AK132230.1 - 5 - 6 1416 89
BC010672.1 - 6 - 7 1465 89
BC021330.1 - 6 - 7 1513 89
BC038028.1 - 6 - 7 836 89
BC055464.1 - 5 - 6 1399 89
BC096029.1 - 6 - 7 1491 89
BC150893.1 - 6 - 7 1460 89
BC157979.1 - 6 - 7 1460 89

View Full Product Details