Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Itgax
Entrez Gene ID
16411
Gene Name
integrin alpha X
Gene Aliases
AI449405, Cd11c, Cr4, N418
Chromosome Location
Chr.7: 128129529 - 128150657 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021334.2 NP_067309.1 24 - 25 2953 79
GenBank mRNA AF211864.1 - 24 - 25 2953 79
AK131133.1 - 24 - 25 2959 79
AK155816.1 - 24 - 25 2975 79
AK170432.1 - 24 - 25 2971 79
BC042840.1 - 24 - 25 2952 79

View Full Product Details