Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Itgax
Entrez Gene ID
16411
Gene Name
integrin alpha X
Gene Aliases
AI449405, Cd11c, Cr4, N418
Chromosome Location
Chr.7: 128129529 - 128150657 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021334.2 NP_067309.1 9 - 10 1071 81
GenBank mRNA AF211864.1 - 9 - 10 1071 81
AK131133.1 - 9 - 10 1077 81
AK155816.1 - 9 - 10 1093 81
AK170432.1 - 9 - 10 1089 81
BC042840.1 - 9 - 10 1070 81

View Full Product Details