Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim30a
Entrez Gene ID
20128
Gene Name
tripartite motif-containing 30A
Gene Aliases
Rpt-1, Rpt1, Trim30
Chromosome Location
Chr.7: 104409025 - 104465193 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009099.2 NP_033125.2 5 - 6 991 88
GenBank mRNA AF220014.1 - 5 - 6 908 88
AK134144.1 - 5 - 6 960 88
AK137506.1 - 5 - 6 992 88
J03776.1 - 5 - 6 908 88

View Full Product Details