Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plau
Entrez Gene ID
18792
Gene Name
plasminogen activator, urokinase
Gene Aliases
u-PA, uPA
Chromosome Location
Chr.14: 20836662 - 20843388 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008873.3 NP_032899.1 9 - 10 1074 72
GenBank mRNA BC120709.1 - 8 - 9 1014 72
BC120713.1 - 8 - 9 1014 72
X02389.1 - 9 - 10 1029 72

View Full Product Details