Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Prkcb
- Entrez Gene ID
-
18751
- Gene Name
- protein kinase C, beta
- Gene Aliases
-
A130082F03Rik, PKC-Beta, Pkcb, Prkcb1, Prkcb2
- Chromosome Location
-
Chr.7: 122289078 - 122634402 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
X59274.1 |
- |
16 - 17 |
1903 |
88 |
View Full Product Details