The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028765.3 | NP_083041.1 | 6 - 7 | 832 | 84 |
GenBank mRNA | AK004775.1 | - | 6 - 7 | 834 | 84 |