Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam53b
Entrez Gene ID
77938
Gene Name
family with sequence similarity 53, member B
Gene Aliases
A930008G19Rik, mKIAA0140
Chromosome Location
Chr.7: 132712078 - 132813196 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175268.4 NP_780477.2 4 - 5 1144 71
NM_212473.1 NP_997638.1 4 - 5 1207 71
GenBank mRNA AK031576.1 - 3 - 4 988 71
AK051299.1 - 4 - 5 1208 71
AK147350.1 - 4 - 5 1540 71
AK147725.1 - 4 - 5 1336 71
BC035523.1 - 4 - 5 1139 71

View Full Product Details